tert-Butyl (1-bromo-2-oxo-6,9,12-trioxa-3-azatetradecan-14-yl)carbamate 95%

Supplier: Ambeed
Warning

A756030-100MG A756030-1G A756030-250MG A756030-5G
76652-578EA 34 USD
76652-578 76652-580 76652-582 76652-584
tert-Butyl (1-bromo-2-oxo-6,9,12-trioxa-3-azatetradecan-14-yl)carbamate 95%
tert-Butyl (1-bromo-2-oxo-6,9,12-trioxa-3-azatetradecan-14-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₉BrN₂O₆
MDL Number: MFCD23378554
CAS Number: 1421933-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR