Bromo-PEG2-CH2CO2tBu 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(2-(2-bromoethoxy)ethoxy)acetate, 2-Methyl-2-propanyl [2-(2-bromoethoxy)ethoxy]acetate, Bromo-PEG2-CH2-Boc

A755925-250MG A755925-5G A755925-1G
76652-574EA 34 USD
76652-574 76652-576 77598-986
Bromo-PEG2-CH2CO2tBu 95%
Bromo-PEG2-CH2CO2tBu

Precaución: for Research Use Only

Formula: C₁₀H₁₉BrO₄
MDL Number: MFCD28122957
CAS Number: 1807518-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR