Propargyl-PEG3-amine 97%

Supplier: Ambeed
Warning

A755212-1G A755212-250MG A755212-100MG A755212-25G A755212-5G
76652-560EA 81 USD
76652-560 76652-562 77598-976 77622-786 77783-930
Propargyl-PEG3-amine 97%
Propargyl-PEG3-amine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MDL Number: MFCD20720810
CAS Number: 932741-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR