2,2-Dimethyl-4-oxo-3,7,10,13-tetraoxahexadecan-16-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: Acid-PEG3-C2-Boc

A754902-100MG A754902-1G A754902-250MG
76652-554EA 195.21 USD
76652-554 76652-556 76652-558
2,2-Dimethyl-4-oxo-3,7,10,13-tetraoxahexadecan-16-oic acid 95%
2,2-Dimethyl-4-oxo-3,7,10,13-tetraoxahexadecan-16-oic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₆O₇
MW: 306.35 g/mol
MDL Number: MFCD28015771
CAS Number: 1807539-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR