tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Bromoacetamido-C2-PEG2-NH-Boc, Bromoacetamido-​PEG2-​Boc-​amine

A754804-1G A754804-5G
76652-552EA 65 USD
76652-552 77783-918
tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₅BrN₂O₅
MDL Number: MFCD28142467
CAS Number: 182244-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR