1-Bromo-3,6,9,12-tetraoxapentadec-14-yne 95%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG4-Br

A754783-1G A754783-5G
76652-550EA 90 USD
76652-550 77622-770
1-Bromo-3,6,9,12-tetraoxapentadec-14-yne 95%
1-Bromo-3,6,9,12-tetraoxapentadec-14-yne

Precaución: for Research Use Only

Formula: C₁₁H₁₉BrO₄
MDL Number: MFCD28950764
CAS Number: 1308299-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR