Amino-PEG8-alcohol 95%

Supplier: Ambeed
Warning

Synonyms: Amino-dPEG®8-alcohol, NH2-PEG8-OH, 23-Amino-3,6,9,12,15,18,21-heptaoxatricosan-1-ol

A754680-100MG A754680-250MG A754680-1G A754680-5G
76652-546EA 27 USD
76652-546 76652-548 77598-954 77622-768
Amino-PEG8-alcohol 95%
Amino-PEG8-alcohol

Precaución: for Research Use Only

Formula: C₁₆H₃₅NO₈
MDL Number: MFCD11041128
CAS Number: 352439-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR