Propargyl-PEG8-t-butyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propargyl-PEG8-Boc
A754424-1G
A754424-250MG
A754424-100MG
76652-544EA
98
USD
76652-544
77622-760
77783-915
Propargyl-PEG8-t-butyl ester 95%
Propargyl-PEG8-t-butyl ester
Precaución: for Research Use Only
Formula:
C₂₄H₄₄O₁₀ MW: 492.6 g/mol |
MDL Number:
MFCD28976700 CAS Number: 2055014-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...