Amino-PEG7-alcohol 98%

Supplier: Ambeed
Warning

Synonyms: NH2-PEG7, 20-Amino-3,6,9,12,15,18-hexaoxaicosan-1-ol

A754377-1G A754377-5G A754377-250MG
76652-542EA 80 USD
76652-542 77569-440 77622-758
Amino-PEG7-alcohol 98%
Amino-PEG7-alcohol

Precaución: For research use only.

Formula: C₁₄H₃₁NO₇
MDL Number: MFCD28385479
CAS Number: 1425973-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR