Propargyl-PEG6-acid 95%

Supplier: Ambeed
Warning

A754239-1G A754239-250MG A754239-100MG A754239-5G
76652-538EA 320 USD
76652-538 77598-938 77622-752 77622-754
Propargyl-PEG6-acid 95%
Propargyl-PEG6-acid

Precaución: for Research Use Only

Formula: C₁₆H₂₈O₈
MW: 348.389 g/mol
MDL Number: MFCD28950778
CAS Number: 1951438-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR