Amino-PEG7-Amine 95%

Supplier: Ambeed
Warning

Synonyms: 3,6,9,12,15,18,21-Heptaoxatricosane-1,23-diamine

A754234-100MG A754234-250MG
76652-534EA 92.3 USD
76652-534 76652-536
Amino-PEG7-Amine 95%
Amino-PEG7-Amine

Precaución: for Research Use Only

Formula: C₁₆H₃₆N₂O₇
MW: 368.466 g/mol
MDL Number: MFCD09752883
CAS Number: 332941-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR