Cbz-N-amido-PEG2-acid 95%

Supplier: Ambeed
Warning

Synonyms: Cbz-NH-PEG2-C2-acid, Cbz-n-amido-peg2-acid

A753944-250MG A753944-100MG A753944-1G A753944-5G
76652-514EA 35 USD
76652-514 77622-738 77622-740 77783-904
Cbz-N-amido-PEG2-acid 95%
Cbz-N-amido-PEG2-acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₆
MDL Number: MFCD28142483
CAS Number: 1347750-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR