Biotin-PEG4-amine 95%

Supplier: Ambeed
Warning

Synonyms: N-(14-Amino-3,6,9,12-tetraoxatetradecyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Amine-PEG4-Biotin, N-Biotinyl-3,6,9,12-tetraoxatetradecane-1,14-diamine, N-[2-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide

A753942-5G A753942-100MG A753942-250MG
76652-512EA 611 USD
76652-512 77598-928 77598-932
Biotin-PEG4-amine 95%
Biotin-PEG4-amine

Precaución: for Research Use Only

Formula: C₂₀H₃₈N₄O₆S
MDL Number: MFCD26142992
CAS Number: 663171-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR