Amyltriethylammonium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: Amyltriethylammonium Bis(trifluoromethanesulphonyl)imide, Triethylpentylammonium Bis(trifluoromethanesulphonyl)imide, N,N,N-triethylpentan-1-aminium bis((trifluoromethyl)sulphonyl)amide, Triethylpentylammonium Bis(trifluoromethanesulfonyl)imide

A753826-1G A753826-25G A753826-5G
76652-504EA 13 USD
76652-504 76652-506 76652-508
Amyltriethylammonium bis(trifluoromethanesulfonyl)imide 98%
Amyltriethylammonium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₃H₂₆F₆N₂O₄S₂
MW: 452.47 g/mol
MDL Number: MFCD15072143
CAS Number: 906478-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR