2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene 95%

Supplier: Ambeed
Warning

A753813-100MG A753813-250MG
76652-500EA 57.29 USD
76652-500 76652-502
2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene 95%
2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene

Precaución: for Research Use Only

Formula: C₂₆H₁₂Br₄
MW: 644.00 g/mol
MDL Number: MFCD22581445
CAS Number: 27192-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR