Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

A753559-1G A753559-25G A753559-5G
76652-476EA 10 USD
76652-476 76652-478 76652-480
Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulfonyl)imide 98%
Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₇H₃₄F₆NO₅PS₂
MW: 541.55 g/mol
MDL Number: MFCD11112190
CAS Number: 959698-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR