1,5-Naphthalenebis(trifluoromethanesulfonate) 95%

Supplier: Ambeed
Danger

Synonyms: 1,5-NAphthalenebis(trifluoromethanesulphonate), 1,5-Naphthaleneditriflate

A753523-10G A753523-1G A753523-5G
76652-470EA 194 USD
76652-470 76652-472 76652-474
1,5-Naphthalenebis(trifluoromethanesulfonate) 95%
1,5-Naphthalenebis(trifluoromethanesulfonate)

Precaución: for Research Use Only

Formula: C₁₂H₆F₆O₆S₂
MDL Number: MFCD23380211
CAS Number: 152873-79-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR