1,3,5-Tricyclohexylbromobenzene 95%

Supplier: Ambeed
Warning

A753414-100MG A753414-250MG A753414-25G A753414-5G
76652-466EA 12 USD
76652-466 76652-468 77783-897 77783-898
1,3,5-Tricyclohexylbromobenzene 95%
1,3,5-Tricyclohexylbromobenzene

Precaución: for Research Use Only

Formula: C₂₄H₃₅Br
MDL Number: MFCD22370173
CAS Number: 97443-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR