Tributylmethylphosphonium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: TRibutylmethylphosphonium bis(trifluoromethanesulphonyl)imide

A753399-1G A753399-25G A753399-5G
76652-460EA 21 USD
76652-460 76652-462 76652-464
Tributylmethylphosphonium bis(trifluoromethanesulfonyl)imide 98%
Tributylmethylphosphonium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₅H₃₀F₆NO₄PS₂
MW: 497.49 g/mol
MDL Number: MFCD15072200
CAS Number: 324575-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR