S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-((Dimethylcarbamothioyl)thio)acetic acid, S-(N,N-Dimethylthiocarbamoyl)mercaptoacetic Acid

A752813-1G A752813-25G A752813-5G
76652-442EA 24 USD
76652-442 76652-444 76652-446
S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid 95%
S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₂S₂
MW: 179.25 g/mol
MDL Number: MFCD00021765
CAS Number: 4007-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR