N-(2-Carboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam 95%

Supplier: Ambeed
Warning

Synonyms: N-(2-CArboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam

A752677-1G A752677-250MG
76652-438EA 264.17 USD
76652-438 76652-440
N-(2-Carboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam 95%
N-(2-Carboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam

Precaución: for Research Use Only

Formula: C₁₈H₁₉Cl₂NO₅S
MW: 432.31 g/mol
MDL Number: MFCD04117914
CAS Number: 193202-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR