2,3,5,6-Tetra(9H-carbazol-9-yl)terephthalonitrile 97%

Supplier: Ambeed
Warning

A752563-100MG A752563-1G A752563-250MG A752563-50MG
76652-420EA 82.76 USD
76652-420 76652-422 76652-424 77618-078
2,3,5,6-Tetra(9H-carbazol-9-yl)terephthalonitrile 97%
2,3,5,6-Tetra(9H-carbazol-9-yl)terephthalonitrile

Precaución: for Research Use Only

Formula: C₅₆H₃₂N₆
MW: 788.89 g/mol
MDL Number: MFCD28369266
CAS Number: 1416881-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR