N-Fmoc-L-alanine monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-L-alanine monohydrate, N-(9-Fluorenylmethoxycarbonyl)-L-alanine monohydrate, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine monohydrate, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected monohydrate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine monohydrate, (-)-L-Alanine, N-FMOC protected monohydrate, (-)-FMOC-L-Ala-OH monohydrate, Fmoc-Ala-OH·H₂O, FMoc-ala-oh h2o, Fmoc-Ala-OH monohydrate

A752490-100G A752490-25G A752490-5G
76652-412EA 51 USD
76652-412 76652-414 76652-416
N-Fmoc-L-alanine monohydrate 98%
N-Fmoc-L-alanine monohydrate

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄·H₂O
MDL Number: MFCD00037139
CAS Number: 207291-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR