Myristic anhydride 95%

Supplier: Ambeed
Warning

Synonyms: Myristic anhydride

A752176-25G A752176-5G A752176-1G
76652-404EA 140 USD
76652-404 76652-406 77622-708
Myristic anhydride 95%
Myristic anhydride

Precaución: for Research Use Only

Formula: C₂₈H₅₄O₃
MDL Number: MFCD00056151
CAS Number: 626-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR