Diethyl-4,4'-azodibenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Azobenzene-4,4'-dicarboxylic Acid Diethyl Ester, Diethyl 4,4'-azodibenzoate, 4,4'-Azodibenzoic Acid Diethyl Ester, Diethyl Azobenzene-4,4'-dicarboxylate

A752148-100MG A752148-1G A752148-250MG A752148-5G
76652-396EA 24.41 USD
76652-396 76652-398 76652-400 76652-402
Diethyl-4,4'-azodibenzoate 98%
Diethyl-4,4'-azodibenzoate

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₄
MW: 326.35 g/mol
MDL Number: MFCD00096305
CAS Number: 7250-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR