2-Isopropoxyethyl methanesulfonate 95%

Supplier: Ambeed
Danger

Synonyms: MEthanesulphonic acid 2-isopropoxyethyl ester, Methanesulphonic acid 2-Isopropoxyethyl Ester

A752054-1G A752054-250MG A752054-5G
76652-384EA 54.11 USD
76652-384 76652-386 76652-388
2-Isopropoxyethyl methanesulfonate 95%
2-Isopropoxyethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₆H₁₄O₄S
MW: 182.23 g/mol
MDL Number: MFCD02093478
CAS Number: 235097-76-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR