Fmoc-NH-PEG8-CH2COOH 95%

Supplier: Ambeed
Warning

A751633-100MG A751633-250MG A751633-5G A751633-1G A751633-25G
76652-378EA 54.11 USD
76652-378 76652-380 77552-296 77573-408 77573-410
Fmoc-NH-PEG8-CH2COOH 95%
Fmoc-NH-PEG8-CH2COOH

Precaución: for Research Use Only

Formula: C₃₃H₄₇NO₁₂
MW: 649.726 g/mol
MDL Number: MFCD27635163
CAS Number: 868594-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR