Biotin-PEG3-acid 95%

Supplier: Ambeed
Warning

Synonyms: 14-Oxo-18-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10-trioxa-13-azaoctadecanoic acid

A751592-100MG A751592-250MG A751592-5G
76652-370EA 45 USD
76652-370 76652-372 77622-692
Biotin-PEG3-acid 95%
Biotin-PEG3-acid

Precaución: for Research Use Only

Formula: C₁₉H₃₃N₃O₇S
MDL Number: MFCD22574817
CAS Number: 252881-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR