2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azahexadecan-16-yl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

A751534-1G A751534-250MG A751534-100MG A751534-25G A751534-5G
76652-366EA 71 USD
76652-366 76652-368 77598-902 77622-688 77622-690
2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azahexadecan-16-yl 4-methylbenzenesulfonate 95%
2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azahexadecan-16-yl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₀H₃₃NO₈S
MDL Number: MFCD27635161
CAS Number: 1246999-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR