Bromo-PEG3-t-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Br-PEG3-C2-Boc

A750957-100MG A750957-1G A750957-250MG A750957-5G A750957-25G
76652-350EA 40.32 USD
76652-350 76652-352 76652-354 77598-896 77622-682
Bromo-PEG3-t-butyl ester 95%
Bromo-PEG3-t-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₅BrO₅
MW: 341.239 g/mol
MDL Number: MFCD22574821
CAS Number: 782475-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR