2-(Tritylthio)ethanamine 98%

Supplier: Ambeed
Warning

A750815-1G A750815-250MG A750815-25G A750815-5G
76652-344EA 55.17 USD
76652-344 76652-346 76652-348 77552-294
2-(Tritylthio)ethanamine 98%
2-(Tritylthio)ethanamine

Precaución: for Research Use Only

Formula: C₂₁H₂₁NS
MW: 319.4631 g/mol
MDL Number: MFCD00822629
CAS Number: 1095-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR