3,6,9,12,15,18-Hexaoxahenicos-20-yn-1-ol 95%

Supplier: Ambeed
Warning

A750759-250MG A750759-5G A750759-1G
76652-338EA 61 USD
76652-338 77569-424 77598-892
3,6,9,12,15,18-Hexaoxahenicos-20-yn-1-ol 95%
3,6,9,12,15,18-Hexaoxahenicos-20-yn-1-ol

Precaución: For research use only.

Formula: C₁₅H₂₈O₇
MDL Number: MFCD22574806
CAS Number: 944560-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR