Hydroxy-PEG3-acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy)propanoic acid

A750553-100MG A750553-250MG A750553-1G
76652-328EA 29 USD
76652-328 76652-330 77573-404
Hydroxy-PEG3-acid 95%
Hydroxy-PEG3-acid

Precaución: for Research Use Only

Formula: C₉H₁₈O₆
MDL Number: MFCD26793743
CAS Number: 518044-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR