Hydroxy-PEG3-ethyl ester 95%

Supplier: Ambeed
Warning

A750523-1G A750523-250MG
76652-324EA 135.8 USD
76652-324 76652-326
Hydroxy-PEG3-ethyl ester 95%
Hydroxy-PEG3-ethyl ester

Precaución: for Research Use Only

Formula: C₉H₁₈O₅
MW: 206.236 g/mol
MDL Number: MFCD22683283
CAS Number: 1379649-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR