Methyl 2-thioxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A750366-100MG A750366-250MG
76652-312EA 16 USD
76652-312 76652-314
Methyl 2-thioxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylate 98%
Methyl 2-thioxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MDL Number: MFCD09192253
CAS Number: 64375-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR