m-PEG9-acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,5,8,11,14,17,20,23,26-Nonaoxanonacosan-29-oic acid

A750267-1G A750267-250MG
76652-308EA 152.77 USD
76652-308 76652-310
m-PEG9-acid 95%
m-PEG9-acid

Precaución: for Research Use Only

Formula: C₂₀H₄₀O₁₁
MW: 456.525 g/mol
MDL Number: MFCD27977514
CAS Number: 2576495-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR