Propargyl-PEG5-amine 95%

Supplier: Ambeed
Warning

Synonyms: 3,6,9,12,15-Pentaoxaoctadec-17-yn-1-amine

A750249-100MG A750249-1G A750249-250MG
76652-302EA 19 USD
76652-302 76652-304 76652-306
Propargyl-PEG5-amine 95%
Propargyl-PEG5-amine

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₅
MW: 275.341 g/mol
MDL Number: MFCD27977505
CAS Number: 1589522-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR