Hydroxy-PEG8-t-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-hydroxy-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate, Hydroxy-PEG8-Boc, Hydroxy-dPEG®8-t-butyl ester

A748984-100MG A748984-1G A748984-250MG
76652-294EA 61.54 USD
76652-294 76652-296 76652-298
Hydroxy-PEG8-t-butyl ester 95%
Hydroxy-PEG8-t-butyl ester

Precaución: for Research Use Only

Formula: C₂₃H₄₆O₁₁
MW: 498.60 g/mol
MDL Number: MFCD11041105
CAS Number: 1334177-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR