N,N-Dipropylethylamine 98%

Supplier: Ambeed
Danger

A748975-1G A748975-25G A748975-5G
76652-288EA 15 USD
76652-288 76652-290 76652-292
N,N-Dipropylethylamine 98%
N,N-Dipropylethylamine

Precaución: for Research Use Only

Formula: C₈H₁₉N
MDL Number: MFCD00048685
CAS Number: 20634-92-8
UN: 2733
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR