2-Octenylsuccinic anhydride (cis and trans mixture) 98%

Supplier: Ambeed
Warning

A748863-1000G A748863-100G A748863-25G A748863-500G
76652-280EA 275 USD
76652-280 76652-282 76652-284 76652-286
2-Octenylsuccinic anhydride (cis and trans mixture) 98%
2-Octenylsuccinic anhydride (cis and trans mixture)

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₃
MDL Number: MFCD00047133
CAS Number: 42482-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR