Ethyl 4-ethyl-3-oxohexanoate 95%
Supplier: Ambeed
|
Warning
|
A748675-100MG
A748675-250MG
A748675-1G
A748675-5G
76652-276EA
25
USD
76652-276
76652-278
77749-505
77749-506
Ethyl 4-ethyl-3-oxohexanoate 95%
Ethyl 4-ethyl-3-oxohexanoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈O₃ |
MDL Number:
MFCD19269441 CAS Number: 91007-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...