1-Butyl-3-methylimidazolium dicyanamide 98%

Supplier: Ambeed
Warning

A748573-1G A748573-25G A748573-5G A748573-100G
76652-270EA 6 USD
76652-270 76652-272 76652-274 77783-853
1-Butyl-3-methylimidazolium dicyanamide 98%
1-Butyl-3-methylimidazolium dicyanamide

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₅
MDL Number: MFCD07784457
CAS Number: 448245-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR