Mal-amido-PEG4-acid 98%

Supplier: Ambeed
Warning

Synonyms: 19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic acid, 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic acid

A748445-100MG A748445-250MG
76652-266EA 48 USD
76652-266 76652-268
Mal-amido-PEG4-acid 98%
Mal-amido-PEG4-acid

Precaución: for Research Use Only

Formula: C₁₈H₂₈N₂O₉
MDL Number: MFCD13184954
CAS Number: 1263045-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR