4-(1,2,2-Triphenylvinyl)benzaldehyde 97%

Supplier: Ambeed
Warning

A748299-100MG A748299-1G A748299-250MG A748299-5G
76652-258EA 6.37 USD
76652-258 76652-260 76652-262 76652-264
4-(1,2,2-Triphenylvinyl)benzaldehyde 97%
4-(1,2,2-Triphenylvinyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₂₇H₂₀O
MW: 360.4471 g/mol
MDL Number: MFCD31619164
CAS Number: 1289218-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR