Ro 10-5824 2HCl 98%

Supplier: Ambeed
Warning

A747660-100MG A747660-10MG A747660-1MG A747660-25MG A747660-50MG A747660-5MG
76652-234EA 215 USD
76652-234 76652-236 76652-238 76652-240 76652-242 76652-244
Ro 10-5824 2HCl 98%
Ro 10-5824 2HCl

Precaución: for Research Use Only

Formula: C₁₇H₂₂Cl₂N₄
MDL Number: MFCD08703115
CAS Number: 189744-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR