(R)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-D-Dab(Boc)-OMe·HCl

A747243-1G A747243-250MG
76652-230EA 122.01 USD
76652-230 76652-232
(R)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochloride 95%
(R)-Methyl 2-amino-4-((tert-butoxycarbonyl)amino)butanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₄
MW: 268.7377 g/mol
MDL Number: MFCD08275859
CAS Number: 1052649-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR