n-Hexadecyl-β-D-maltoside 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Hexadecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, n-Hexadecyl β-D-maltopyranoside

A746904-100MG A746904-1G A746904-250MG
76652-216EA 46 USD
76652-216 76652-218 76652-220
n-Hexadecyl-β-D-maltoside 98%
n-Hexadecyl-β-D-maltoside

Precaución: for Research Use Only

Formula: C₂₈H₅₄O₁₁
MDL Number: MFCD01310880
CAS Number: 98064-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR