(S)-Methyl 6-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Lys(Z)-OMe

A746900-10G A746900-1G A746900-5G A746900-25G
76652-210EA 73 USD
76652-210 76652-212 76652-214 77622-660
(S)-Methyl 6-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate 97%
(S)-Methyl 6-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate

Precaución: for Research Use Only

Formula: C₂₀H₃₀N₂O₆
MDL Number: MFCD03788041
CAS Number: 73548-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR