3-Benzyl-5-((2-nitrophenoxy)methyl)dihydrofuran-2(3H)-one 98%

Supplier: Ambeed
Warning

A745543-10MG A745543-1MG A745543-25MG A745543-5MG
76652-176EA 89 USD
76652-176 76652-178 76652-180 76652-182
3-Benzyl-5-((2-nitrophenoxy)methyl)dihydrofuran-2(3H)-one 98%
3-Benzyl-5-((2-nitrophenoxy)methyl)dihydrofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₅
MDL Number: MFCD30187363
CAS Number: 890405-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR