1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one 95%

Supplier: Ambeed
Warning

A743266-10MG A743266-1MG A743266-5MG A743266-50MG
76652-166EA 51 USD
76652-166 76652-168 76652-170 77618-044
1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one 95%
1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one

Precaución: for Research Use Only

Formula: C₁₇H₁₅ClN₆O₂
MDL Number: MFCD31579854
CAS Number: 1883711-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR